Learn More
Aat Bioquest mBBr [Monobromobimane] *CAS 71418-44-5*
SDP
Supplier:  Aat Bioquest 633
Encompass_Preferred
mBBr is one of the most popular thiol-reactive fluorescent tags, and is widely used to detect various thiol-containing biomolecules such as glutathione in cells. It is fluorogenic upon reacting with thiol-containing molecules.
Catalog No. 50-195-2941