Learn More
Medchemexpress LLC MBCQ | 10MG
SDP
Catalog No. 5000227209
Encompass_Preferred
MBCQ is a potent and selective cGMP-specific phosphodiesterase (PDE V PDE5) inhibitor with an IC50 of 19 nM MBCQ lacks inhibitory activity toward other PDE isozymes (all IC50s 100 M) MBCQ dilates coronary arteries via specific inhibition of cGMP-PDE[1][2][3]
Catalog No. 50-002-27209 Supplier Medchemexpress LLC Supplier No. HY11467210MG
MBCQ is a potent and selective cGMP-specific phosphodiesterase (PDE V PDE5) inhibitor with an IC50 of 19 nM MBCQ lacks inhibitory activity toward other PDE isozymes (all IC50s 100 M) MBCQ dilates coronary arteries via specific inhibition of cGMP-PDE[1][2][3]