Learn More
Medchemexpress LLC MBCQ 50mg | 150450-53-6 | 50 MG
SDP
Catalog No. 5000229605
Encompass_Preferred
MBCQ is a selective cGMP-specific phosphodiesterase type 5 (PDE5) inhibitor supplied for research use. It exhibits potent PDE5 inhibition (IC50 19 nM) with minimal activity on other PDE isozymes, and is provided as a solid research reagent suitable for biochemical assays and in vivo studies.
  • Potent PDE5 inhibition (IC50 19 nM).
  • High selectivity versus other PDE isozymes (IC50 >100 μM).
  • High purity suitable for research (99.8%).
  • Soluble in DMSO at 100 mg/mL for in vitro applications.
  • Stable as a powder at -20°C for long-term storage.
  • Available in small pack sizes for laboratory use.
Catalog No. 50-002-29605 Supplier Medchemexpress LLC Supplier No. HY11467250MG
MBCQ is a selective cGMP-specific phosphodiesterase type 5 (PDE5) inhibitor supplied for research use. It exhibits potent PDE5 inhibition (IC50 19 nM) with minimal activity on other PDE isozymes, and is provided as a solid research reagent suitable for biochemical assays and in vivo studies.
  • Potent PDE5 inhibition (IC50 19 nM).
  • High selectivity versus other PDE isozymes (IC50 >100 μM).
  • High purity suitable for research (99.8%).
  • Soluble in DMSO at 100 mg/mL for in vitro applications.
  • Stable as a powder at -20°C for long-term storage.
  • Available in small pack sizes for laboratory use.