Learn More
Medchemexpress LLC HY-114672 5mg Medchemexpress, MBCQ CAS:150450-53-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1146725MG
Encompass_Preferred
Medchemexpress, HY-114672 5mg MBCQ CAS:150450-53-6 MBCQ is a potent and selective cGMP-specific phosphodiesterase (PDE V; PDE5) inhibitor with an IC50 of 19 nM. MBCQ lacks inhibitory activity toward other PDE isozymes (all IC50s>100 μM). MBCQ dilates coronary arteries via specific inhibition of cGMP-PDE. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0874