Learn More
Enzo Life Sciences MBCQ (5mg). CAS: 150450-53-6
SDP
Supplier:  Enzo Life Sciences BMLPD1490005
Encompass_Preferred
Selective inhibitor of cGMP-specific phosphodiesterase (PDE V, IC50=19 nM). Purity: ≥98%. Solubility: Soluble in DMSO. Long Term Storage: Ambient.
Catalog No. 50-201-1381