Learn More
Medchemexpress LLC MBD (7-(p-methoxybenzylamino)-4-nitrobenz-2,1,3-oxadiazole) | 33984-50-8 | 99.6% | C14H12N4O4 | 100 MG
SDP
Catalog No. 5000354624
Encompass_Preferred
MBD is a fluorescent probe utilized for investigating protein and nucleoprotein conformation. It has been successfully applied to bacterial ribosomes as well as bovine trypsinogen and trypsin.
  • Fluorescent probe for investigating protein and nucleoprotein conformation.
  • Successfully applied to bacterial ribosomes, bovine trypsinogen, and trypsin.
  • Strong fluorescence when binding to hydrophobic regions of macromolecules.
Catalog No. 50-003-54624 Supplier Medchemexpress LLC Supplier No. HY141667100MG
MBD is a fluorescent probe utilized for investigating protein and nucleoprotein conformation. It has been successfully applied to bacterial ribosomes as well as bovine trypsinogen and trypsin.
  • Fluorescent probe for investigating protein and nucleoprotein conformation.
  • Successfully applied to bacterial ribosomes, bovine trypsinogen, and trypsin.
  • Strong fluorescence when binding to hydrophobic regions of macromolecules.