Learn More
Aviva Systems Biology Corp MBD1 Antibody - middle region (ARP40172_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40172_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MBD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MBD1. Alternate names: CXXC3, PCM1, RFTApplicationWB. Species reactivity: Dog, Human
Catalog No. 50-014-57316