Learn More
Aviva Systems Biology Corp MBD2 Antibody - N-terminal region (ARP32485_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32485_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MBD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MBD2. Alternate names: DKFZp586O0821, DMTase, NY-CO-41ApplicationWB, IHC. Species reactivity: Human, Mouse, Rat
Catalog No. 50-014-54619