Learn More
ACROBiosystems ACROBiosystems Human MBL Protein, His Tag
Catalog No. 501956925
Encompass_Preferred
Recombinant Protein;1MG;Human MBL, His Tag (MBL-H5220) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Ile 248 (Accession # AAH96179).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.9 kDa. The protein migrates as 30-33 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-195-6925 Supplier ACROBiosystems Supplier No. MBLH52201MG
Recombinant Protein;1MG;Human MBL, His Tag (MBL-H5220) is expressed from human 293 cells (HEK293). It contains AA Glu 21 - Ile 248 (Accession # AAH96179).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.9 kDa. The protein migrates as 30-33 kDa under reducing (R) condition (SDS-PAGE).