Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents QP10091MA20UG
Encompass_Preferred
Product Type: Recombinant Protein. Conjugate/Tag: Flag . Host: Mammal. Species Reactivity: Bovine. Protein Expression: A DNA sequence encoding the Bos taurus (Bovine) Mannose-binding protein C, was expressed in the hosts and tags indicated. AA Sequence: DTETENCENI RKTCPVIACG PPGINGIPGK DGRDGAKGEK GEPGQGLRGS QGPPGKMGPQ GTPGIPGIPG PIGQKGDPGE NMGDYIRLAT SERATLQSEL NQIKNWLIFS LGKRVGKKAF FTNGKKMPFN EVKTLCAQFQ GRVATPMNAE ENRALKDLVT EEAFLGITDQ ETEGKFVDLT GKGVTYQNWN DGEPNNASPG EHCVTLLSDG TWNDIACSAS FLTVCEFSL. Buffer: Tris-based buffer, 50% glycerol. Purity: Greater than 90% as determined by SDS-PAGE. Endotoxin: Not determined. Gene Symbol: MBL. Gene ID: 281297. Uniprot: O02659.
Catalog No. 50-188-5492