Learn More
Aviva Systems Biology Corp MBL2 Antibody - middle region (ARP45676_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45676_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MBL2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MBL2. Alternate names: COLEC1, HSMBPC, MBL, MBP, MBP1, MGC116832, MGC116833, MBL2D, MBP-CApplicationWB. Species reactivity: Cow, Goat, Horse, Human, Pig, Rat, Sheep
Catalog No. 50-285-214