Learn More
Medchemexpress LLC MBL2/COLEC1 Mouse 10ug
SDP
Catalog No. 5000184425
Encompass_Preferred
Recombinant mouse Mannose-binding lectin 2 (MBL2/COLEC1) produced in HEK293 mammalian cells and supplied as a purified protein for use in biochemical and immunoassay applications. The product is intended for researchers performing binding studies, ELISA, and structure-function analyses where mammalian expression and post-translational modifications support native-like protein behavior.
  • Recombinant mouse MBL2 produced in HEK293 cells for native-like folding.
  • Suitable for ELISA, binding assays, and structure-function studies.
  • Purified protein format simplifies assay setup.
  • Mammalian expression supports relevant glycosylation patterns.
  • Supplied as a 10 μg aliquot for small-scale experiments.
Catalog No. 50-001-84425 Supplier Medchemexpress LLC Supplier No. HYP7067410UG
Recombinant mouse Mannose-binding lectin 2 (MBL2/COLEC1) produced in HEK293 mammalian cells and supplied as a purified protein for use in biochemical and immunoassay applications. The product is intended for researchers performing binding studies, ELISA, and structure-function analyses where mammalian expression and post-translational modifications support native-like protein behavior.
  • Recombinant mouse MBL2 produced in HEK293 cells for native-like folding.
  • Suitable for ELISA, binding assays, and structure-function studies.
  • Purified protein format simplifies assay setup.
  • Mammalian expression supports relevant glycosylation patterns.
  • Supplied as a 10 μg aliquot for small-scale experiments.