Learn More
TARGETMOL CHEMICALS INC MBM-55 5MG
SDP
Catalog No. 502581498
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MBM-55 is an effective inhibitor of NIMA related kinase 2 (NEK2) with an IC50 of 1 nM. MBM-55 effectively inhibits the proliferation of cancer cells by inducing cell cycle arrest and apoptosis. purity: 99%
Catalog No. 50-258-1498 Supplier TARGETMOL CHEMICALS INC Supplier No. T119605MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MBM-55 is an effective inhibitor of NIMA related kinase 2 (NEK2) with an IC50 of 1 nM. MBM-55 effectively inhibits the proliferation of cancer cells by inducing cell cycle arrest and apoptosis. purity: 99%