Learn More
Aviva Systems Biology Corp MBNL2 Antibody - middle region (ARP51632_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51632_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MBNL2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MBNL2. Alternate names: DKFZp781H1296, MBLL, MBLL39, MGC120625, MGC120626, MGC120628, PRO2032, RP11-128N14.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60573