Learn More
Aviva Systems Biology Corp MBOAT1 Antibody - N-terminal region (ARP48548_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48548_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MBOAT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MBOAT1. Alternate names: MGC44669, OACT1, dJ434O11.1, LPEAT1ApplicationWB. Species reactivity: Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59855