Learn More
New England Biolabs, Inc. MboI – 2500 units
SDP
Catalog No. 50812026
Encompass
A restriction endonuclease that recognizes the sequence ^GATC_.
Catalog No. 50-812-026 Supplier New England Biolabs, Inc. Supplier No. R0147L
A restriction endonuclease that recognizes the sequence ^GATC_.