Learn More
New England Biolabs, Inc. MboI – 2500 units
SDP
Catalog No. 50812027
Encompass
A restriction endonuclease that recognizes the sequence ^GATC_.
Catalog No. 50-812-027 Supplier New England Biolabs, Inc. Supplier No. R0147M
A restriction endonuclease that recognizes the sequence ^GATC_.