Learn More
New England Biolabs, Inc. MboII – 1500 units
SDP
Catalog No. 50812029
Encompass
A restriction endonuclease that recognizes the sequence GAAGANNNNNNN_N^.
Catalog No. 50-812-029 Supplier New England Biolabs, Inc. Supplier No. R0148L
A restriction endonuclease that recognizes the sequence GAAGANNNNNNN_N^.