Learn More
New England Biolabs, Inc. MboII – 300 units
SDP
Catalog No. 50812030
Encompass
A restriction endonuclease that recognizes the sequence GAAGANNNNNNN_N^.
Catalog No. 50-812-030 Supplier New England Biolabs, Inc. Supplier No. R0148S
A restriction endonuclease that recognizes the sequence GAAGANNNNNNN_N^.