Learn More
Bellco Glass, Inc. mBox Stain Rack PP 1x3"" Slide""
SDP
Catalog No. 501945831
Encompass_Preferred
mBox is designed to immerse 20 1x3" slides in liquid during blocking, denaturing and post-arraying. They also prevent damage during shipping and processing. Designed for easy removal of slides and fits standard micro-tier plate format in centrifuge.
Catalog No. 50-194-5831 Supplier Bellco Glass, Inc. Supplier No. 563830100
mBox is designed to immerse 20 1x3" slides in liquid during blocking, denaturing and post-arraying. They also prevent damage during shipping and processing. Designed for easy removal of slides and fits standard micro-tier plate format in centrifuge.