Learn More
AnaSpec This is fragment of the myelin basic protein MBP which corresponds to amino acids 84-96 of the murine sequence 86-98 in guinea pig 87-99 in human. VHFFKNIVTPRTP
Supplier:  AnaSpec AS29211
Encompass
This is fragment of the myelin basic protein MBP which corresponds to amino acids 84-96 of the murine sequence 86-98 in guinea pig 87-99 in human. VHFFKNIVTPRTP
Catalog No. NC3715299