Learn More
Medchemexpress LLC MBP Agarose (Dextrin) 6FF | 5 ML
SDP
Catalog No. 5000430193
Encompass_Preferred
MCE MBP Agarose (Dextrin) 6FF is prepared by covalently coupling dextrin to an agarose matrix for one-step purification of fusion proteins. It features high binding capacity, excellent specificity, and superior ligand stability. Bound fusion protein can be gently eluted using 10 mM maltose, preserving biological activity. The tag can be removed by site-specific protease cleavage.
  • High binding capacity
  • Excellent specificity
  • Superior ligand stability
  • Enables one-step purification of fusion proteins
  • Gentle elution preserves biological activity
  • Enhances solubility of heterologous proteins
  • Improves expression levels
  • Promotes proper protein folding
  • Tag removable by specific proteases
  • Suitable for protein expression, localization, purification, and immunoassays
Catalog No. 50-004-30193 Supplier Medchemexpress LLC Supplier No. HYK02525ML
MCE MBP Agarose (Dextrin) 6FF is prepared by covalently coupling dextrin to an agarose matrix for one-step purification of fusion proteins. It features high binding capacity, excellent specificity, and superior ligand stability. Bound fusion protein can be gently eluted using 10 mM maltose, preserving biological activity. The tag can be removed by site-specific protease cleavage.
  • High binding capacity
  • Excellent specificity
  • Superior ligand stability
  • Enables one-step purification of fusion proteins
  • Gentle elution preserves biological activity
  • Enhances solubility of heterologous proteins
  • Improves expression levels
  • Promotes proper protein folding
  • Tag removable by specific proteases
  • Suitable for protein expression, localization, purification, and immunoassays