Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MBP-MAPK-Substrate | 138028-00-9 | 99.6% | 967.09 | 5 MG
SDP

Catalog No. 5000424251
Encompass_Preferred

MBP MAPK Substrate is utilized as an exogenous substrate for MAPK. It is intended for research use only, and not for sale to patients.

  • Used as an exogenous substrate for MAPK
  • Appearance: solid
  • Color: white to off-white
  • Sequence: Ala-Pro-Arg-Thr-Pro-Gly-Gly-Arg-R
  • Sequence shortening: APRTPGGRR
  • Target: p38 MAPK
  • Pathway: MAPK/ERK pathway

Catalog No. 50-004-24251 Supplier Medchemexpress LLC Supplier No. HYP24565MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MBP MAPK Substrate is utilized as an exogenous substrate for MAPK. It is intended for research use only, and not for sale to patients.

  • Used as an exogenous substrate for MAPK
  • Appearance: solid
  • Color: white to off-white
  • Sequence: Ala-Pro-Arg-Thr-Pro-Gly-Gly-Arg-R
  • Sequence shortening: APRTPGGRR
  • Target: p38 MAPK
  • Pathway: MAPK/ERK pathway

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.