Learn More
Proteintech Group Inc MBP-Trap Agarose beads
SDP
Catalog No. 502258526 Shop All Proteintech Group Inc Products
Encompass
The ChromoTek MBP-Trap Agarose are affinity beads for IP of MBP-fusion proteins. They comprise MBP Nanobodies/ VHHs coupled to agarose beads.
This item is not returnable. View return policy
Catalog No. 50-225-8526 Supplier Proteintech Group Inc Supplier No. MBTA20
The ChromoTek MBP-Trap Agarose are affinity beads for IP of MBP-fusion proteins. They comprise MBP Nanobodies/ VHHs coupled to agarose beads.