Learn More
Medchemexpress LLC HY-101525 5mg Medchemexpress, MBP146-78 CAS:188343-77-3 Purity:>98%
SDP
Catalog No. 501932158
Encompass_Preferred
Medchemexpress, HY-101525 5mg MBP146-78 CAS:188343-77-3 MBP146-78 is a potent and selective inhibitor of cGMP dependent protein kinases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2158 Supplier Medchemexpress LLC Supplier No. HY1015255MG
Medchemexpress, HY-101525 5mg MBP146-78 CAS:188343-77-3 MBP146-78 is a potent and selective inhibitor of cGMP dependent protein kinases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.