Learn More
TARGETMOL CHEMICALS INC MBQ-167 10MG
SDP
Catalog No. 502877527
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. MBQ-167 is a dual inhibitor of Rac/Cdc42 (IC50s 103 nM for Rac 1/2/3 and 78 nM for Cdc42 in MDA-MB-231 cells, respectively). Purity 98.74%
Catalog No. 50-287-7527 Supplier TARGETMOL CHEMICALS INC Supplier No. T1602110MG
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. MBQ-167 is a dual inhibitor of Rac/Cdc42 (IC50s 103 nM for Rac 1/2/3 and 78 nM for Cdc42 in MDA-MB-231 cells, respectively). Purity 98.74%