Learn More
Medchemexpress LLC HY-112842 10mg Medchemexpress, MBQ-167 CAS:2097938-73-1 Purity:>98%
SDP
Catalog No. 501969188
Encompass_Preferred
Medchemexpress, HY-112842 10mg MBQ-167 CAS:2097938-73-1 MBQ-167 is a dual Rac/Cdc42 inhibitor, with IC50s of 103 nM for Rac 1/2/3 and 78 nM for Cdc42 in MDA-MB-231 cells, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9188 Supplier Medchemexpress LLC Supplier No. HY11284210MG
Medchemexpress, HY-112842 10mg MBQ-167 CAS:2097938-73-1 MBQ-167 is a dual Rac/Cdc42 inhibitor, with IC50s of 103 nM for Rac 1/2/3 and 78 nM for Cdc42 in MDA-MB-231 cells, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.