Learn More
Neta Scientific Agilent Vial,MicroV,snap,1.5ml,ambr,100PK
SDP
Catalog No. 50929351
Encompass_Preferred
This item is manufactured by Agilent
Agilent Vial, amber, microsampling, wine-glass shape, 12 x 32 snap top,15 uL at the bottom taper, for small sample volume or high recovery,100/pk. Vial size: 12 x 32 mm (11 mm cap) - Micro-V Vials
UNSPSC Code  41121806
- 58
Catalog No. 50-929-351 Supplier Neta Scientific Supplier No. 51843556
This item is manufactured by Agilent
Agilent Vial, amber, microsampling, wine-glass shape, 12 x 32 snap top,15 uL at the bottom taper, for small sample volume or high recovery,100/pk. Vial size: 12 x 32 mm (11 mm cap) - Micro-V Vials
UNSPSC Code  41121806
- 58