Learn More
Bioworld MBS, 100 mg
Supplier:  Bioworld 218100051
Encompass_Preferred
Specifications:HeterobifunctionalReactive groups maleimide and NHS esterPrimary amine and sulhydryl reactiveHapten-carrier protein conjugation 1,2.Soluble in DMSO and chloroformStorage: Shipped at ambient temperature, upon receipt store at 4°C.References: 1.) 1.) Lerner, R.A., et al. (1981) PNAS (USA) 78, 3403-3407. 2.) Chamberlain, N.R., et al. (1989) Infection and Immunity 57, 2878-2885. 3.) Dell' Arciprete, L., et al. (1988) J. Immunol. 140, 2466-2471.
Catalog No. 50-199-254