Learn More
Bruker Daltonics MBT GALAXY HCCA MATRIX, 7.5 mg, pre-portioned powder, for MALDI-TOF MS applications, high-purity α-Cyano-4-hydroxycinnamic acid
Catalog No. NC1169104
Encompass_Preferred
The Bruker Scientific LLC 1823405 MBT Galaxy HCCA Matrix is a high-purity α-Cyano-4-hydroxycinnamic acid (HCCA) matrix optimized for MALDI-TOF MS applications. This pre-portioned matrix simplifies the preparation of MALDI solutions, crucial for accurate measurements of peptides, proteins, and microorganism identification. It integrates seamlessly with Bruker's MALDI Biotyper systems and the MBT Galaxy automated workflow, ensuring reproducible results and a streamlined laboratory process.
  • High-purity matrix for clean spectra
  • Pre-portioned vials for simplified preparation
  • Automated, contact-free matrix deposition
  • Minimized contamination risks during use
  • Stability for up to 1 week at 20-25°C
  • Storage conditions of 2-8°C when unopened
  • Compatible with Bruker MALDI Biotyper systems
Catalog No. NC1169104 Supplier Bruker Daltonics Supplier No. 1823405
The Bruker Scientific LLC 1823405 MBT Galaxy HCCA Matrix is a high-purity α-Cyano-4-hydroxycinnamic acid (HCCA) matrix optimized for MALDI-TOF MS applications. This pre-portioned matrix simplifies the preparation of MALDI solutions, crucial for accurate measurements of peptides, proteins, and microorganism identification. It integrates seamlessly with Bruker's MALDI Biotyper systems and the MBT Galaxy automated workflow, ensuring reproducible results and a streamlined laboratory process.
  • High-purity matrix for clean spectra
  • Pre-portioned vials for simplified preparation
  • Automated, contact-free matrix deposition
  • Minimized contamination risks during use
  • Stability for up to 1 week at 20-25°C
  • Storage conditions of 2-8°C when unopened
  • Compatible with Bruker MALDI Biotyper systems