Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProteoSpec™ MBTFA, Ricca Chemical
SDP

Catalog No. MB122041012 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
10 x 1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MB122041012 10 x 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MB122041012 Supplier Ricca Chemical Company Supplier No. RMB1220410X1C2

GC Derivatization Reagent

Chemical Identifiers

CAS 685-27-8
Molecular Formula C5H3F6NO2
Molecular Weight (g/mol) 223.074
InChI Key AWGBWLXGUPTXHF-UHFFFAOYSA-N
PubChem CID 69635
IUPAC Name 2,2,2-trifluoro-N-methyl-N-(2,2,2-trifluoroacetyl)acetamide
SMILES CN(C(=O)C(F)(F)F)C(=O)C(F)(F)F

Specifications

Packaging Glass Ampule
CAS Min % 100
CAS Max % 100
Density 1.547
Flash Point 45°C
Physical Form Liquid
Color Colorless
Boiling Point 121°C
Solubility Miscible with water
Quantity 10 x 1 g
Formula Weight 223.07
Percent Purity 98.0% min.
Grade Molecular Biology
Assay GC
Chemical Name or Material ProteoSpec™ MBTFA N-Methyl-bis(trifluoroacetamide)
Show More Show Less
ShelfLife 36 months
Recommended Storage Ambient Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.