Learn More
Specifications
Specifications
| Chemical Name or Material | MBTH Indicator |
| Name Note | 0.05% Aqueous, for Cyanide |
| CAS | 149022-15-1, 7732-18-5 |
| CAS Max % | 99.95, 0.05 |
| Identification | Passes Test |
| Packaging | Poly Bottle |
| Sensitivity | Passes Test |
| Specific Gravity | 1g/mL |
| Linear Formula | C8H9N3S·HCl |
| Decomposition Information | Nitrogen oxides; Carbon monoxide; Carbon dioxide; Sulfur compounds; Hydrogen chloride |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.