Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MBTH Indicator, 0.05% Aqueous, for Cyanide, Certified, LabChem™

Catalog No. LC163307 Shop All LabChem Products
Change view
Click to view available options
Quantity:
125 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC163307 125 mL
LC163301 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC163307 Supplier LabChem, Inc. Supplier No. LC163307
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material MBTH Indicator
Name Note 0.05% Aqueous, for Cyanide
CAS 149022-15-1, 7732-18-5
CAS Max % 99.95, 0.05
Identification Passes Test
Packaging Poly Bottle
Sensitivity Passes Test
Specific Gravity 1g/mL
Linear Formula C8H9N3S·HCl
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide; Sulfur compounds; Hydrogen chloride
Density 1g/mL
Color Colorless to Brown
Solubility Information Soluble in water
Quantity 125 mL
Physical Form Liquid
Formula Weight 215.7
Grade Certified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.