Learn More
Hach Company MBTH Powder Pillows, pk/100
Supplier:  Hach Company 2257169
Encompass
Reagent for low range formaldehyde determination by the MBTH photometric method. Hach Method 8110 Pack of 100 Powder Pillows
Catalog No. 50-199-5649