Learn More
Medchemexpress LLC HY-112565 5mg Medchemexpress, MBX-4132 CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1125655MG
Encompass_Preferred
Medchemexpress, HY-112565 5mg MBX-4132 CAS: MBX-4132, a member of a chemical class called oxadiazoles that inhibit trans translation by binding to the bacterial ribosome. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9105