Learn More
Cayman Chemical MBX-8025sodIum salt 10mg
SDP
Catalog No. 502938749
Encompass_Preferred
A PPARδ agonist; >750- and >2,500-fold selective for PPARδ over PPARα and PPARγ; reduces increases in fasting blood glucose and serum insulin levels, and decreases insulin resistance in Alms1 mutant (foz/foz) mice fed an atherogenic diet as a model of diet-induced obesity, type 2 diabetes, and NASH at 10 mg/kg per day for eight weeks; decreases serum ALT, as well as serum and hepatic cholesterol and triglyceride, levels and reduces markers of NASH in the same model
Catalog No. 50-293-8749 Supplier Cayman Chemical Supplier No. 2876110 MG
A PPARδ agonist; >750- and >2,500-fold selective for PPARδ over PPARα and PPARγ; reduces increases in fasting blood glucose and serum insulin levels, and decreases insulin resistance in Alms1 mutant (foz/foz) mice fed an atherogenic diet as a model of diet-induced obesity, type 2 diabetes, and NASH at 10 mg/kg per day for eight weeks; decreases serum ALT, as well as serum and hepatic cholesterol and triglyceride, levels and reduces markers of NASH in the same model