Learn More
Alkali Scientific MC-24™ High Speed Microcentrifuge with COMBI-Rotor, 115v
SDP
Catalog No. 502335939
Encompass_Preferred
MC-24 Touch is the first microcentrifuge in its class to feature touch screen control. In addition to the next generation control panel, the centrifuge also offers high speed (up to 13,500 rpm/16,800 xg) and a unique COMBI-Rotor that accepts both standard microtubes (1.5/2.0mL) and PCR tubes/strips (0.2mL), making this the perfect partner for any molecular biology laboratory. The large, color touch screen shows actual speed (in rpm or rcf) along with set and remaining time. Up to 9 speed/time profiles can be stored in memory and the home screen displays a “mySpin” icon for instant recalls a designated favorite profile.
Catalog No. 50-233-5939 Supplier Alkali Scientific Supplier No. RC2417
MC-24 Touch is the first microcentrifuge in its class to feature touch screen control. In addition to the next generation control panel, the centrifuge also offers high speed (up to 13,500 rpm/16,800 xg) and a unique COMBI-Rotor that accepts both standard microtubes (1.5/2.0mL) and PCR tubes/strips (0.2mL), making this the perfect partner for any molecular biology laboratory. The large, color touch screen shows actual speed (in rpm or rcf) along with set and remaining time. Up to 9 speed/time profiles can be stored in memory and the home screen displays a “mySpin” icon for instant recalls a designated favorite profile.