Learn More
Alkali Scientific Alkali Scientific™ MC-24R™ (C2417-R-NR) Refrigerated High Speed Microcentrifuge w/o rotor
SDP
Catalog No. 503029648
Encompass_Preferred
The Alkali Scientific™ C2417-R-NR-E MC-24R refrigerated microcentrifuge features a next generation touch control panel, high speed centrifugation (up to 13,500 rpm / 16,800 xg). Refrigerated High Speed Microcentrifuge 230V Rotor not included Touch operation Status indicator light Powerful motor drive provides speeds up to 13,500 rmp (16,800xg) Powerful refrigeration, as low as -20°C
Catalog No. 50-302-9648 Supplier Alkali Scientific Supplier No. RC2417RNR
The Alkali Scientific™ C2417-R-NR-E MC-24R refrigerated microcentrifuge features a next generation touch control panel, high speed centrifugation (up to 13,500 rpm / 16,800 xg). Refrigerated High Speed Microcentrifuge 230V Rotor not included Touch operation Status indicator light Powerful motor drive provides speeds up to 13,500 rmp (16,800xg) Powerful refrigeration, as low as -20°C