Learn More
Alkali Scientific MC-24R C2417R Refrigerated Hig
SDP
Catalog No. 503029597
Encompass_Preferred
Alkali Scientifics Fagship MC-24R (C2417R) refrigerated microcentrifuge features a next generation touch control panel high speed centrifugation(up to 13500 rpm / 17157 xg) and a unique COMBIRotorthat accepts both standard microtubes (1 5/2 0 ml) and PCR tubes/strips (0 2 ml) eliminating the need for a dedicated PCR strip/tube rotor These features combine to make this the perfect partner for any molecular biology laboratory The MC-24R (C2417R) has available packages that include Alkalis unique COMBI-Rotoras well as access to an optional high speed spin column rotor reaching 17157 xg a hermetically sealed rotor and a four-place PCR strip rotor The MC-24R (C2417R) features adjustable acceleration/decelerations rates real time rpm and rcf conversion program storage up to 99 and a unique status indicator light The status indicator light informs the user if the unit is running in standby or has completed a run The MC-24R is a compact and convenient refrigerated centrifuge perfectly sui
Catalog No. 50-302-9597 Supplier Alkali Scientific Supplier No. RS2417R
Add to Cart
Add to Cart
Request a Quote
Alkali Scientifics Fagship MC-24R (C2417R) refrigerated microcentrifuge features a next generation touch control panel high speed centrifugation(up to 13500 rpm / 17157 xg) and a unique COMBIRotorthat accepts both standard microtubes (1 5/2 0 ml) and PCR tubes/strips (0 2 ml) eliminating the need for a dedicated PCR strip/tube rotor These features combine to make this the perfect partner for any molecular biology laboratory The MC-24R (C2417R) has available packages that include Alkalis unique COMBI-Rotoras well as access to an optional high speed spin column rotor reaching 17157 xg a hermetically sealed rotor and a four-place PCR strip rotor The MC-24R (C2417R) features adjustable acceleration/decelerations rates real time rpm and rcf conversion program storage up to 99 and a unique status indicator light The status indicator light informs the user if the unit is running in standby or has completed a run The MC-24R is a compact and convenient refrigerated centrifuge perfectly sui