Learn More
Bellco Glass, Inc. MicroCarrier Flask Complete,36L
SDP
Supplier:  Bellco Glass, Inc. 196536000
Encompass_Preferred
MICROCARRIER SPINNER FLASKS-PROMOTES OPTIMAL GROWTH OF SUSPENSION AND MICROCARRIER CULTURE-IMPELLER HEIGHT ADJUSTS THROUGH COMPRESSION FITTING-INDENTED BOTTOM (UP TO 1L) PREVENTS ACCUMULATION OF MICROCARRIER BEADS UNDER IMPELLEROD X OAH:410 X 545 (with cap)CAP:100MMSIDEARM:45MM. MADE IN THE USA.
Catalog No. 50-194-5020