Learn More
Bellco Glass, Inc. MicroCarrier Flask Complete,500mL,70mm
SDP
Catalog No. 501944969
Encompass_Preferred
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. Size: 500ml Center Cap:70mm Sidearm cap: 45mm
Catalog No. 50-194-4969 Supplier Bellco Glass, Inc. Supplier No. 196500500
Promotes optimal growth of suspension and micro-carrier culture. Impeller height adjust through compression fitting center cap. The indented bottom prevents accumulation of micro-carrier beads under impeller. MADE IN THE USA. Size: 500ml Center Cap:70mm Sidearm cap: 45mm