Learn More
Medchemexpress LLC MC-GGFG-(7ethanol-10NH2-11F-camptothecin) | 2987937-38-0 | 95.6% | 922.91 g·mol⁻¹ | C46H47FN8O12 | 1 MG
SDP
Catalog No. 5000204361
Encompass_Preferred
MC-GGFG-(7ethanol-10NH2-11F-Camptothecin) is a drug-linker conjugate intended for antibody-drug conjugate (ADC) research. It features a protease-cleavable MC-GGFG linker attached to a camptothecin derivative and is supplied in small mg-scale quantities.
  • Protease-cleavable linker for ADC conjugation.
  • High purity of 95.6%.
  • Available in small mg-scale pack sizes.
  • Molecular weight 922.91 g·mol⁻¹ and formula C46H47FN8O12.
  • Intended for research use.
Catalog No. 50-002-04361 Supplier Medchemexpress LLC Supplier No. HY1565151MG
MC-GGFG-(7ethanol-10NH2-11F-Camptothecin) is a drug-linker conjugate intended for antibody-drug conjugate (ADC) research. It features a protease-cleavable MC-GGFG linker attached to a camptothecin derivative and is supplied in small mg-scale quantities.
  • Protease-cleavable linker for ADC conjugation.
  • High purity of 95.6%.
  • Available in small mg-scale pack sizes.
  • Molecular weight 922.91 g·mol⁻¹ and formula C46H47FN8O12.
  • Intended for research use.