Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MC-GGFG-(7ethanol-10NH2-11F-camptothecin) | 2987937-38-0 | 95.6% | 922.91 g/mol | C46H47FN8O12 | 5 MG
SDP

Catalog No. 5000204459
Encompass_Preferred

A protease-cleavable drug-linker conjugate composed of a GGFG peptide linker attached to a camptothecin derivative (7-ethanol-10-amino-11-fluoro-camptothecin). Supplied in small milligram quantities for antibody-drug conjugate research; characterized by reported purity, molecular weight, formula, CAS number, solubility in DMSO, and storage recommendations.

  • Protease-cleavable GGFG peptide linker
  • Camptothecin derivative payload with fluorine substitution
  • Reported purity of 95.6%
  • Soluble in DMSO (110 mg/mL; ultrasonic recommended)
  • Store at -20°C and protect from light; in solution store at -80°C (6 months) or -20°C (1 month)
  • Provided in small milligram pack sizes

Catalog No. 50-002-04459 Supplier Medchemexpress LLC Supplier No. HY1565155MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A protease-cleavable drug-linker conjugate composed of a GGFG peptide linker attached to a camptothecin derivative (7-ethanol-10-amino-11-fluoro-camptothecin). Supplied in small milligram quantities for antibody-drug conjugate research; characterized by reported purity, molecular weight, formula, CAS number, solubility in DMSO, and storage recommendations.

  • Protease-cleavable GGFG peptide linker
  • Camptothecin derivative payload with fluorine substitution
  • Reported purity of 95.6%
  • Soluble in DMSO (110 mg/mL; ultrasonic recommended)
  • Store at -20°C and protect from light; in solution store at -80°C (6 months) or -20°C (1 month)
  • Provided in small milligram pack sizes

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.