Learn More
Medchemexpress LLC HY-114233 10mg , MC-GGFG-Exatecan CAS:1600418-29-8 Purity:>98%
SDP
Catalog No. 502259459
Encompass_Preferred
HY-114233 10mg MC-GGFG-Exatecan CAS: 1600418-29-8 MC-GGFG-Exatecan (MC-GGFG-DX8951) is a drug-linker conjugate for ADC. MC-GGFG-Exatecan is a DX8951 (a DNA topoisomerase I inhibitor) derivative with protease cleavable MC-GGFG linker. MC-GGFG-Exatecan shows antitumor activity and can be used to prepare DX8951 antibody conjugate (ADC). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9459 Supplier Medchemexpress LLC Supplier No. HY11423310MG
HY-114233 10mg MC-GGFG-Exatecan CAS: 1600418-29-8 MC-GGFG-Exatecan (MC-GGFG-DX8951) is a drug-linker conjugate for ADC. MC-GGFG-Exatecan is a DX8951 (a DNA topoisomerase I inhibitor) derivative with protease cleavable MC-GGFG linker. MC-GGFG-Exatecan shows antitumor activity and can be used to prepare DX8951 antibody conjugate (ADC). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.