Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MC-GGFG-NH-CH2-O-CH2-(s-cyclopropane)-COOH | 2626930-84-3 | 97.4% | 656.68 g/mol | C31H40N6O10 | 25 MG
SDP

Catalog No. 5000203884
Encompass_Preferred

MC-GGFG-NH-CH2-O-CH2-(s-cyclopropane)-COOH is an ADC linker compound used for conjugation in antibody-drug conjugate research and bioconjugation workflows. It is supplied as a high-purity small molecule suitable for linker incorporation and payload attachment in ADC development.

  • High purity (97.4%).
  • Molecular formula C31H40N6O10.
  • Molecular weight 656.68 g/mol.
  • Supplied in milligram quantities for research use.
  • Documentation available: data sheet, certificate of analysis, and safety data sheet.

Catalog No. 50-002-03884 Supplier Medchemexpress LLC Supplier No. HYW73108525MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MC-GGFG-NH-CH2-O-CH2-(s-cyclopropane)-COOH is an ADC linker compound used for conjugation in antibody-drug conjugate research and bioconjugation workflows. It is supplied as a high-purity small molecule suitable for linker incorporation and payload attachment in ADC development.

  • High purity (97.4%).
  • Molecular formula C31H40N6O10.
  • Molecular weight 656.68 g/mol.
  • Supplied in milligram quantities for research use.
  • Documentation available: data sheet, certificate of analysis, and safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.