Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mc-ggfg-nh-ch2-o-ch2-(s-cyclopropane)-cooh | 2626930-84-3 | 97.4% | 656.68 g/mol | C31H40N6O10 | 5 MG
SDP

Catalog No. 5000203886
Encompass_Preferred

MC-GGFG-NH-CH2-O-CH2-(s-cyclopropane)-COOH is an antibody-drug conjugate (ADC) linker provided as a camptothecin-conjugated drug-linker for ADC research and conjugation studies.

  • Provided as a camptothecin-conjugated drug-linker for ADC development.
  • Reported purity of 97.4%.
  • Molecular weight 656.68 g/mol.
  • Molecular formula C31H40N6O10.
  • Supplied in small research pack sizes suitable for conjugation workflows.
  • Intended for research use only and not for human clinical use.

Catalog No. 50-002-03886 Supplier Medchemexpress LLC Supplier No. HYW7310855MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MC-GGFG-NH-CH2-O-CH2-(s-cyclopropane)-COOH is an antibody-drug conjugate (ADC) linker provided as a camptothecin-conjugated drug-linker for ADC research and conjugation studies.

  • Provided as a camptothecin-conjugated drug-linker for ADC development.
  • Reported purity of 97.4%.
  • Molecular weight 656.68 g/mol.
  • Molecular formula C31H40N6O10.
  • Supplied in small research pack sizes suitable for conjugation workflows.
  • Intended for research use only and not for human clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.