Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mc-gly-gly-d-phe | 472.49 g/mol | C23H28N4O7 | 25 MG
SDP

Catalog No. 5000203524
Encompass_Preferred

MC-Gly-Gly-D-Phe is a cleavable peptide linker used in the synthesis of antibody-drug conjugates (ADCs). It contains a maleimide group and a Gly-Gly-D-Phe peptide sequence that enables enzymatic cleavage to release payloads in target cells.

  • Cleavable peptide linker for ADC synthesis
  • Protease-sensitive Gly-Gly-D-Phe sequence
  • Solid, white to off-white appearance
  • Molecular weight 472.49 g/mol, formula C23H28N4O7
  • Available in small vial sizes (5 mg-100 mg)
  • Recommended storage at -20 °C

Catalog No. 50-002-03524 Supplier Medchemexpress LLC Supplier No. HY44235A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MC-Gly-Gly-D-Phe is a cleavable peptide linker used in the synthesis of antibody-drug conjugates (ADCs). It contains a maleimide group and a Gly-Gly-D-Phe peptide sequence that enables enzymatic cleavage to release payloads in target cells.

  • Cleavable peptide linker for ADC synthesis
  • Protease-sensitive Gly-Gly-D-Phe sequence
  • Solid, white to off-white appearance
  • Molecular weight 472.49 g/mol, formula C23H28N4O7
  • Available in small vial sizes (5 mg-100 mg)
  • Recommended storage at -20 °C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.