Learn More
Medchemexpress LLC MC-Gly-Gly-Phe-Gly-C | 10mg
SDP
Catalog No. 5000202249
Encompass_Preferred
MC-Gly-Gly-Phe-Gly-Cyclobutanecarboxylic-Exatecan is a agent-linker conjugate for ADC contains the ADC linker (MC-Gly-Gly-Phe-Gly-Cyclobutanecarboxylic acid) and a DNA topoisomerase I inhibitor Exatecan (HY-13631) MC-Gly-Gly-Phe-Gly-Cyclobutanecarboxylic-Exatecan synthesized ADC can be used in study of cancer[1]
Catalog No. 50-002-02249 Supplier Medchemexpress LLC Supplier No. HY14730710MG
MC-Gly-Gly-Phe-Gly-Cyclobutanecarboxylic-Exatecan is a agent-linker conjugate for ADC contains the ADC linker (MC-Gly-Gly-Phe-Gly-Cyclobutanecarboxylic acid) and a DNA topoisomerase I inhibitor Exatecan (HY-13631) MC-Gly-Gly-Phe-Gly-Cyclobutanecarboxylic-Exatecan synthesized ADC can be used in study of cancer[1]