Learn More
Medchemexpress LLC Mc-gly-gly-phe-gly-pab-oh | 2632342-05-1 | 98.8% | 634.68 g/mol | C32H38N6O8 | 10 MG
SDP
Catalog No. 5000203303
Encompass_Preferred
Mc-Gly-Gly-Phe-Gly-PAB-OH is a cleavable antibody-drug conjugate (ADC) linker containing a Mc cap, a Gly-Gly-Phe-Gly peptide sequence, and a para-aminobenzyl (PAB) spacer terminating in a free carboxylic acid. It is designed to be cleaved under intracellular conditions to release cytotoxic payloads for targeted delivery.
  • Cleavable linker enabling intracellular payload release.
  • Contains a Mc cap and GGFG peptide sequence for stability and cleavage specificity.
  • Para-aminobenzyl spacer provides a self-immolative release mechanism.
  • High purity suitable for research applications.
  • Available in multiple small-scale pack sizes for reagent optimization.
Catalog No. 50-002-03303 Supplier Medchemexpress LLC Supplier No. HY13643210MG
Mc-Gly-Gly-Phe-Gly-PAB-OH is a cleavable antibody-drug conjugate (ADC) linker containing a Mc cap, a Gly-Gly-Phe-Gly peptide sequence, and a para-aminobenzyl (PAB) spacer terminating in a free carboxylic acid. It is designed to be cleaved under intracellular conditions to release cytotoxic payloads for targeted delivery.
  • Cleavable linker enabling intracellular payload release.
  • Contains a Mc cap and GGFG peptide sequence for stability and cleavage specificity.
  • Para-aminobenzyl spacer provides a self-immolative release mechanism.
  • High purity suitable for research applications.
  • Available in multiple small-scale pack sizes for reagent optimization.