Learn More
Medchemexpress LLC Mc-Gly-Gly-Phe-Gly-PAB-OH | 2632342-05-1 | 98.8% | 634.68 g/mol | C32H38N6O8 | 50 MG
SDP
Catalog No. 5000203870
Encompass_Preferred
Mc-Gly-Gly-Phe-Gly-PAB-OH is a cleavable ADC linker containing a para-aminobenzyl self-immolative spacer connected to a Gly-Gly-Phe-Gly peptide sequence; it is designed to release payloads under proteolytic conditions for antibody-drug conjugates. Supplied as a white to yellow solid with recommended low-temperature storage for long-term stability.
  • Cleavable linker with PAB self-immolative spacer.
  • Protease-cleavable peptide sequence for controlled payload release.
  • White to yellow solid physical form.
  • High purity suitable for research applications.
  • Recommended sealed, low-temperature storage to preserve integrity.
Catalog No. 50-002-03870 Supplier Medchemexpress LLC Supplier No. HY13643250MG
Mc-Gly-Gly-Phe-Gly-PAB-OH is a cleavable ADC linker containing a para-aminobenzyl self-immolative spacer connected to a Gly-Gly-Phe-Gly peptide sequence; it is designed to release payloads under proteolytic conditions for antibody-drug conjugates. Supplied as a white to yellow solid with recommended low-temperature storage for long-term stability.
  • Cleavable linker with PAB self-immolative spacer.
  • Protease-cleavable peptide sequence for controlled payload release.
  • White to yellow solid physical form.
  • High purity suitable for research applications.
  • Recommended sealed, low-temperature storage to preserve integrity.